Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2 Rabbit anti-Birds, Bovine, Chicken, Clawed frog, Dog, Fruit fly, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89115480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-480 Supplier Abnova Corporation Supplier No. PAB11971
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Sod2.

Specifications

Antigen SOD2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Sod2.
Dilution Western Blot (0.5 ug/mL) Immunohistochemistry (1-10 ug/mL) Immunoprecipitation (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.0 (50% glycerol, 0.09% sodium azide)
Gene Sod2
Gene Symbols Sod2
Host Species Rabbit
Immunogen Rat Sod2 (Mn Superoxide dismutase).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24787
Test Specificity This antibody detects a 25 KDa protein on SDS-PAGE immunoblots corresponding to the molecular mass of Mn SOD.
Target Species Avian, Bovine, Canine, Chicken, Drosophila, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.