Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2, Rabbit, Clone: SP76, Abnova™

Catalog No. 89108346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-346 Supplier Abnova Corporation Supplier No. MAB10031

Rabbit monoclonal antibody raised against synthetic peptide of SOX2.

This intronless gene encodes a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of this gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. Mutations in this gene have been associated with optic nerve hypoplasia and with syndromic microphthalmia, a severe form of structural eye malformation. This gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). [provided by RefSeq

Specifications

Antigen SOX2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP76
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of SOX2.
Dilution Western Blot (1:25) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene SOX2
Gene Alias ANOP3/MCOPS3/MGC2413
Gene Symbols SOX2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human SOX2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.