Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX8/SOX9/SOX17/SOX18, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-608 Supplier Abnova Corporation Supplier No. PAB26962
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SOX8/SOX9/SOX17/SOX18.

Specifications

Antigen SOX8/SOX9/SOX17/SOX18
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SOX8/SOX9/SOX17/SOX18.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene Alias MGC24837
Gene Symbols SOX8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human SOX8/SOX9/SOX17/SOX18.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30812/6662/64321/54345
Test Specificity SOX8/SOX9/SOX17/SOX18 polyclonal antibody detects endogenous levels of SOX8/SOX9/SOX17/SOX18 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.