Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-Sox9, Alexa Fluor 488, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ AB5535AF488
Description
Specifically detects Sox9 in Chicken, Human, Mouse, Rat samples, and it is validated for Immunocytochemistry
Transcription factor SOX-9 (UniProt P48436; also known as Sex-determining region Y-box 9 protein, SRY-related HMG-box gene 9) is encoded by the SOX9 (also known as CMD1, CMPD1, SRA1) gene (Gene ID 6662) in human. Sox family of genes are related to the mammalian sex determining gene SRY. These genes contain sequences that encode for the HMG-box domains that mediate sequence-specific DNA binding. Sox genes encode transcriptional regulators involved in cell fate decision during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at at least 40 genomic loci and at least 30 Sox genes have been identified.
Specifications
Sox9 | |
Polyclonal | |
Purified rabbit conjugate antibody in PBS with 15mg/mL BSA and 0.1% sodium azide. | |
SOX9 | |
KLH-conjugated linear peptide corresponding to the C-terminus of human Sox9. | |
100 μL | |
Epigenetics and Nuclear Function | |
NP_000337 | |
Stable for one year at 2°C to 8°C from date of receipt. |
Immunocytochemistry | |
Alexa Fluor 488 | |
P48436 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Chicken, Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction