Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAG5 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114821 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-821 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-821 Supplier Abnova Corporation Supplier No. PAB1038

Rabbit polyclonal antibody raised against synthetic peptide of SPAG5.

This gene encodes a protein associated with the mitotic spindle apparatus. The encoded protein may be involved in the functional and dynamic regulation of mitotic spindles. [provided by RefSeq

Specifications

Antigen SPAG5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description sperm associated antigen 5
Formulation In PBS (0.02% sodium azide)
Gene SPAG5
Gene Alias DEEPEST, MAP126, hMAP126
Gene Symbols SPAG5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human SPAG5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10615
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.