Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPDYA, Rabbit, DyLight 549, Polyclonal Antibody, Abnova™

Catalog No. 89117502 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-502 Supplier Abnova Corporation Supplier No. PAB15207
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SPDYA.

Specifications

Antigen SPDYA
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 549
Description Rabbit polyclonal antibody raised against synthetic peptide of SPDYA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene SPDYA
Gene Alias MGC110856/MGC57218/Ringo3/SPDY1/SPY1
Gene Symbols SPDYA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-286 of human SPDYA.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245711
Test Specificity This is specific to Speedy/Ringo protein. This antibody is useful for Western blot analysis, where a band is seen at ∼36 KDa (isoform 2). After a longer exposure, a 31 KDa band can be detected (isoform 1). There is also a non-specific band at ∼58 KDa. This antibody was tested against human and mouse brain samples, but failed to detect proteins of the predicted size in either lysate.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.