Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPP1 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89121038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-038 Supplier Abnova Corporation Supplier No. PAB19510

Rabbit polyclonal antibody raised against partial synthetic peptide of mouse Spp1.

Sequence: DPKSKEDDRYLKFRISHE

Specifications

Antigen SPP1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial synthetic peptide of mouse Spp1.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Gene Spp1
Gene Alias AA960535/AI790405/Apl-1/BNSP/BSPI/Bsp/ETA-1/Eta/OP/Opn/Opnl/Ric/Spp-1/minopontin
Gene Symbols Spp1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 268-285 at C-terminus of mouse Spp1.
Purification Method Immunoaffinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20750
Test Specificity The synthetic peptide of this antibody is identical to the related rat sequences.
Target Species Mouse, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.