Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRYD5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120973 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-973 Supplier Abnova Corporation Supplier No. PAB19438
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SPRYD5.

Specifications

Antigen SPRYD5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SPRYD5.
Dilution Western Blot (0.25-0.5 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene SPRYD5
Gene Alias MGC10977/TRIM51
Gene Symbols SPRYD5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids near C-terminus of human SPRYD5.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84767
Test Specificity SPRYD5 antibody is predicted to not cross-react with other SPRYD protein family members. At least two isoforms of SPRYD5 are known to exist; this antibody will detect both isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.