Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTBN2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89115534 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-534 Supplier Abnova Corporation Supplier No. PAB12023
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant SPTBN2.

Specifications

Antigen SPTBN2
Applications Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant SPTBN2.
Dilution Immunoprecipitation (2-5 ug/mg lysate) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene SPTBN2
Gene Alias GTRAP41/SCA5
Gene Symbols SPTBN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to human SPTBN2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6712
Test Specificity The epitope recognized by this antibody maps to a region between residues 2150 and 2200 of human spectrin, beta, non-erythrocytic 2 (spinocerebellar ataxia 5 ) using the numbering given in entry NP_008877.1
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.