Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sptbn2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117815 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-815 Supplier Abnova Corporation Supplier No. PAB15750
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Sptbn2.

Specifications

Antigen Sptbn2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Sptbn2.
Dilution Immunohistochemistry (1:5) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.05% sodium azide)
Gene Sptbn2
Gene Alias Spnb3
Gene Symbols Sptbn2
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 292 amino acids (Q2097-K2388) of rat Sptbn2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29211
Test Specificity This antibody detects rat SPTBN2 protein.
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 200μL distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.