Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-SPTLC1, Polyclonal,

Catalog No. ABS1642 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1642 Supplier MilliporeSigma™ Supplier No. ABS1642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects SPTLC1 in Human samples, and it is validated for Western Blotting

Serine palmitoyltransferase 1 (EC 2.3.1.50; UniProt O15269; also known as LCB 1, Long chain base biosynthesis protein 1, Serine-palmitoyl-CoA transferase 1, SPT 1, SPT1) is encoded by the SPTLC1 (also known as HSAN1, HSAN1A, LCB1) gene (Gene ID 10558) in human. Serine palmitoyltransferase (SPT) catalyzes pyridoxal 5-phosphate-/PLP-dependent condensation of L-serine with palmitoyl-CoA to generate 3-ketodihydrosphingosine, the rate-limiting step for the de novo synthesis of sphingolipids. SPT belongs to the POAS (PLP-dependent α-oxoamine synthase) family of enzymes, members of this family also include 5-amino-levulinic acid synthase, KBL (2-amino-3 ketobutyrate ligase) and AONS (8-amino-7-oxononanoate synthase). Unlike the other known POAS family members that all exist as soluble homodimers, SPT is bound to the outer membrane of the endoplasmic reticulum (ER) as a multisubunit complex composed of three distinct subunits, SPTLC1, SPTLC2 and SPTLC3. SPTLC1 is a 473-amino acid sigle-transmembrane (a.a. 16-36) protein with a short N-terminal lumenal domain (a.a. 1-15) and a large C-terminal cytoplasmic domain (a.a. 37-473). Alternative splicing generates an isoform with a truncated cytoplasmic domain (D143 switched to E143, a.a. 144-473 truncated).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTLC1
Applications Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal antibody serum with 0.05% sodium azide.
Gene Accession No. O15269
Gene Symbols SPTLC1; HSAN1; HSAN1A; LCB1
Host Species Rabbit
Immunogen Two linear peptides corresponding to sequences from the N- and a C-terminal regions of human SPTLC1 cytoplasmic domain.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_006406
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.