Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-SR-alpha, Polyclonal,

Catalog No. ABS1591 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1591 Supplier MilliporeSigma™ Supplier No. ABS1591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects SR-alpha in Canine, Human, Rat samples, and it is validated for Immunoprecipitation, Western Blotting

Signal recognition particle receptor subunit alpha (UniProt P08240; also known as Docking protein alpha, DP-alpha, SR-alpha, SRalpha) is encoded by the SRPR gene (Gene ID 6734) in human. Secretory proteins typically possess N-terminal hydrophobic signal sequences that direct them to the endoplasmic reticulum (ER) membrane, where they are translocated into the ER lumen for subsequent vesicular transport. Signal recognition particle (SRP) delivers target nascent secretory proteins, while they are still being synthesized, together with the associated ribosome to the ER protein translocon (Sec61 complex) via the interaction with SRP receptor (SR) on the ER membrane. SR is a heterodimer composed of a 70kDa peripheral membrane protein (SR alpha subunit or SRα) and a 30kDa singlel-trans membrane protein (SR beta subunit or SRβ). SRα contains an N-terminal SRX domain that mediates its association with SRβ and a C-terminal region which has high homology to the NG domain of SRP54, including a functional GTPase domain. SRβ also possesses a GTPase domain, which interacts with SRα SRX domain only when SRα is in the GTP bound state. SRα forms the major interaction interface with SRP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR-alpha
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P08240
Gene Symbols SRPR
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human SR-alpha.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003130
Target Species Canine, Human, Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.