Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-826 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-826 Supplier Abnova Corporation Supplier No. PAB10386

Rabbit polyclonal antibody raised against synthetic peptide of SRC.

This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen SRC
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
Formulation In PBS, pH 7.3 (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene SRC
Gene Alias ASV, SRC1, c-SRC, p60-Src
Gene Symbols SRC
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 6-19 of human SRC.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6714
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.