Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBF1 Rabbit anti-Mouse, Pig, Rat, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89117490 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-490 Supplier Abnova Corporation Supplier No. PAB15195
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SREBF1.

This gene encodes a transcription factor that binds to the sterol regulatory element-1 (SRE1), which is a decamer flanking the low density lipoprotein receptor gene and some genes involved in sterol biosynthesis. The protein is synthesized as a precursor that is attached to the nuclear membrane and endoplasmic reticulum. Following cleavage, the mature protein translocates to the nucleus and activates transcription by binding to the SRE1. Sterols inhibit the cleavage of the precursor, and the mature nuclear form is rapidly catabolized, thereby reducing transcription. The protein is a member of the basic helix-loop-helix-leucine zipper (bHLH-Zip) transcription factor family. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen SREBF1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against synthetic peptide of SREBF1.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene SREBF1
Gene Alias SREBP-1c/SREBP1/bHLHd1
Gene Symbols SREBF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 300-400 of human SREBF1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6720
Test Specificity This is specific to SREBP1. By Western blot a band is seen at ∼65KDa.
Target Species Mouse, Pig, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.