Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRY, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126556 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-556 Supplier Abnova Corporation Supplier No. PAB24884
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SRY.

This intronless gene encodes a transcription factor that is a member of the high mobility group (HMG)-box family of DNA-binding proteins. This protein is the testis-determining factor (TDF), which initiates male sex determination. Mutations in this gene give rise to XY females with gonadal dysgenesis (Swyer syndrome); translocation of part of the Y chromosome containing this gene to the X chromosome causes XX male syndrome. [provided by RefSeq

Specifications

Antigen SRY
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SRY.
Dilution Western Blot (1:500-1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In 1X PBS, pH 7.2 (0.05% sodium azide)
Gene SRY
Gene Alias TDF/TDY
Gene Symbols SRY
Host Species Rabbit
Immunogen A synthetic peptide corresponding to SRY.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6736
Test Specificity SRY polyclonal antibody detects endogenous levels of SRY protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.