Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5A Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116091 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-091 Supplier Abnova Corporation Supplier No. PAB12786
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of STAT5A.

Specifications

Antigen STAT5A
Applications ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of STAT5A.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene Alias MGF/STAT5
Gene Symbols STAT5A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding Y694 (DGYVK) of human, mouse and rat STAT5A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6776/20850/24918
Test Specificity This antibody recognizes STAT5A without a phosphorylated sites of Tyr694. It does not cross-react with phosphospecific peptide.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.