Learn More
STEAP2 Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STEAP2.
Supplier: Abnova Corporation PAB1062
Description
This gene is a member of the STEAP family and encodes a multi-pass membrane protein that localizes to the Golgi complex, the plasma membrane, and the vesicular tubular structures in the cytosol. A highly similar protein in mouse has both ferrireductase and cupric reductase activity, and stimulates the cellular uptake of both iron and copper in vitro. Increased transcriptional expression of the human gene is associated with prostate cancer progression. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: ISRKLKRIKKGWEKSQSpecifications
STEAP2 | |
Polyclonal | |
six transmembrane epithelial antigen of the prostate 2 | |
STEAP2 | |
STEAP2 | |
A synthetic peptide corresponding to amino acids 455-467 of human STEAP2. | |
RUO | |
261729 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
IPCA1, PCANAP1, PUMPCn, STAMP1, STMP | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only