Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116513 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-513 Supplier Abnova Corporation Supplier No. PAB13265
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of STIM2.

This gene is a member of the stromal interaction molecule (STIM) family and likely arose, with related family member STIM1, from a common ancestral gene. It encodes a type 1 transmembrane protein whose function is currently unknown, but is a likely adhesion molecule with a role in early hematopoiesis. Alternative translation initiation from an AUG and a non-AUG (UUG) start site results in the production of two different isoforms. [provided by RefSeq

Specifications

Antigen STIM2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of STIM2.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene STIM2
Gene Alias FLJ39527/KIAA1482
Gene Symbols STIM2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 18 amino acids of human STIM2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57620
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.