Learn More
STK11 (phospho S334), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of STK11.
Supplier: Abnova Corporation PAB16956
Description
This gene, which encodes a member of the serine/threonine kinase family, regulates cell polarity and functions as a tumor suppressor. Mutations in this gene have been associated with Peutz-Jeghers syndrome, an autosomal dominant disorder characterized by the growth of polyps in the gastrointestinal tract, pigmented macules on the skin and mouth, and other neoplasms. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Specifications
STK11 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of STK11. | |
In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) | |
LKB1/PJS | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of STK11 only when phosphorylated at serine 334. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
STK11 | |
STK11 | |
Synthetic phosphopeptide corresponding to residues surrounding S334 of human STK11. | |
RUO | |
6794 | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only