Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK17A Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134461 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-461 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-461 Supplier Abnova Corporation Supplier No. PAB9860

Rabbit polyclonal antibody raised against synthetic peptide of STK17A.

This gene is a member of the DAP kinase-related apoptosis-inducing protein kinase family and encodes an autophosphorylated nuclear protein with a protein kinase domain. The protein has apoptosis-inducing activity. [provided by RefSeq

Specifications

Antigen STK17A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description serine/threonine kinase 17a
Formulation In PBS (0.02% sodium azide)
Gene STK17A
Gene Alias DRAK1
Gene Symbols STK17A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 5-19 of human STK17A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9263
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.