Learn More
STK24, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STK24.
Supplier: Abnova Corporation PAB2297
Description
The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases (Zhou et al., 2000 [PubMed 10644707]).[supplied by OMIM
Specifications
STK24 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of STK24. | |
In PBS (0.09% sodium azide) | |
MST-3/MST3/MST3B/STE20/STK3 | |
Rabbit | |
Protein G purification | |
RUO | |
8428 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
STK24 | |
STK24 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human STK24. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only