Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRN Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-526 Supplier Abnova Corporation Supplier No. PAB12016
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of STRN.

Specifications

Antigen STRN
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of STRN.
Dilution Immunofluorescence (1:5000) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.02% sodium azide)
Gene STRN
Gene Alias MGC125642/SG2NA
Gene Symbols STRN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 350-400 of human STRN.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6801
Test Specificity This antibody does not cross-react with SG2NA.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.