Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STT3B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-744 Supplier Abnova Corporation Supplier No. PAB23044
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant STT3B.

The SIMP protein contains a highly immunogenic minor histocompatibility antigen epitope of 9 amino acids, B6(dom1). Like ITM1 (MIM 601134), SIMP is homologous to yeast STT3, an oligosaccharyltransferase essential for cell proliferation (McBride et al., 2002 [PubMed 12439619]).[supplied by OMIM

Sequence: NPPVEDSSDEDDKRNQGNLYDKAGKVRKHATEQEKTEEGLGP

Specifications

Antigen STT3B
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant STT3B.
Dilution Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene STT3B
Gene Alias FLJ90106/SIMP/STT3-B
Gene Symbols STT3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human STT3B.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201595
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.