Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-365 Supplier Abnova Corporation Supplier No. PAB0716
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SUMO1.

This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last four amino acids of the carboxy-terminus have been cleaved off. Several pseudogenes have been reported for this gene. Alternate transcriptional splice variants encoding different isoforms have been characterized. [provided by RefSeq

Sequence: AKPSTEDLGDKKEGEY

Specifications

Antigen SUMO1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SUMO1.
Dilution Immunohistochemistry (1:1000-1:2000) Western Blot (1:2000-1:4000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene SUMO1
Gene Alias DAP-1/GMP1/OFC10/PIC1/SENP2/SMT3/SMT3C/SMT3H3/SUMO-1/UBL1
Gene Symbols SUMO1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 6-21 of human SUMO1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7341
Test Specificity This antiserum recognizes human SUMO-1 and not ubiquitin.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.