Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUP35, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SUP35.
Supplier: Abnova Corporation PAB15578
Specifications
| SUP35 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SUP35. | |
| In antiserum (0.05% sodium azide) | |
| GST1/PNM2/SAL3/SUF12/SUP2/SUP36 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody reacts with S. cerevisiae SUP35. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| SUP35 | |
| SUP35 | |
| A synthetic peptide corresponding to amino acids 494-507 of Saccharomyces cerevisiae SUP35. | |
| RUO | |
| 851752 | |
| Yeast | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction