Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYCP1 Rabbit anti-Mouse, DyLight 549, Polyclonal Antibody, Abnova™

Catalog No. 89117379 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-379 Supplier Abnova Corporation Supplier No. PAB14964
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Sycp1.

Specifications

Antigen SYCP1
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate DyLight 549
Description Rabbit polyclonal antibody raised against synthetic peptide of Sycp1.
Dilution Immunofluorescence (1:100-1:750) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:750) Immunohistochemistry (Frozen sections) (1:750) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene Sycp1
Gene Alias SCP1
Gene Symbols Sycp1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of mouse Sycp1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20957
Test Specificity This is specific to mouse SCP1. This antibody is useful for immunofluorescence and immunohistochemistry (paraffin and frozen sections).
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.