Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYN1 (phospho S9), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134315 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-315 Supplier Abnova Corporation Supplier No. PAB9664
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1.

Specifications

Antigen SYN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene Syn1
Gene Symbols Syn1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S9 of rat Syn1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24949
Test Specificity This antibody also weakly labels the ∼55k synapsin II protein which has a similar phosphorylation site to that of Ser9 on synapsin I.
Target Species Bovine, Frog, Human, Mouse, Rat, Zebrafish
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.