Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYN1 Rabbit anti-Bovine, Polyclonal Antibody, Abnova™

Catalog No. 89117125 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-125 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-125 Supplier Abnova Corporation Supplier No. PAB14534
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native SYN1.

Specifications

Antigen SYN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native SYN1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene SYN1
Gene Alias MGC152080
Gene Symbols SYN1
Host Species Rabbit
Immunogen Native purified bovine SYN1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281510
Target Species Bovine
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.