Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Sync1, Polyclonal,

Catalog No. ABN1006MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1006MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1006MI Supplier MilliporeSigma™ Supplier No. ABN1006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Sync1 in Human, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry

SYNC1 is an intermediate structural protein important in muscle cell function and metabolism. In skeletal muscle, SYNC1 localizes with desmin and Dystrobrevin alpha , and is localized at the myotendinous and neuromuscular junctions, sarcolemma and Z-lines. In myotubes, SYNC1 is detected in a punctate cytoplasmic pattern and is up regulated in various forms of neuromuscular diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sync1
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9H7C4
Gene Symbols SYNC; SYNC1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Sync1 head domain near the N-terminal.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_110413
Target Species Human, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.