Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synuclein (pan), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137917 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-917 Supplier Abnova Corporation Supplier No. PAB14315
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Synuclein (pan).

Sequence: AKEGVVAAAEKTKQGV

Specifications

Antigen Synuclein (pan)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Synuclein (pan).
Dilution Immunohistochemistry (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with diphtheria toxoid) corresponding to human Synuclein (pan).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Overlap specific immunohistochemical staining of alpha-, beta- and gamma synucleins.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.