Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SYP.
Supplier: Abnova Corporation PAB14508
Description
Synaptophysin (p38) is an integral membrane protein of small synaptic vesicles in brain and endocrine cells.[supplied by OMIM
Specifications
SYP | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SYP. | |
In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (1% BSA, 0.05% sodium azide) | |
SYP | |
A synthetic peptide (conjugated with carrier protein) corresponding to C-terminus of human SYP. | |
RUO | |
6855 | |
Store at 4°C. |
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
SYP | |
Rabbit | |
500 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction