Learn More
SYTL5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SYTL5.
Supplier: Abnova Corporation PAB21790
Description
The protein encoded by this gene belongs to the synaptotagmin-like (Slp) protein family, which contains a unique homology domain at the N-terminus, referred to as the Slp homology domain (SHD). The SHD functions as a binding site for Rab27A, which plays a role in protein transport. Expression of this gene is restricted to placenta and liver, suggesting that it might be involved in Rab27A-dependent membrane trafficking in specific tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: KQVNVLGTDVVRQSILRRSPAEEVQSQEQTRQDAEKSDTSPVAGKKASHDGPKRKGFLLSKFRSATRGEIITPKTDTGRSYSLDLDGQHFRSLKSPPGSDRGSTGSSDLNDQEPSpecifications
| SYTL5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SYTL5. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| slp5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 94122 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SYTL5 | |
| SYTL5 | |
| Recombinant protein corresponding to amino acids of human SYTL5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only