Learn More
TAAR6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TAAR6.
Supplier: Abnova Corporation PAB27697
Description
The TAAR6 (TRAR4) gene belongs to the trace amine receptor family. Trace amines are endogenous amine compounds that are chemically similar to classic biogenic amines like dopamine, norepinephrine, serotonin, and histamine. Trace amines were thought to be 'false transmitters' that displace classic biogenic amines from their storage and act on transporters in a fashion similar to the amphetamines, but the identification of brain receptors specific to trace amines indicates that they also have effects of their own (Duan et al., 2004 [PubMed 15329799]).[supplied by OMIM
Specifications
TAAR6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TAAR6. | |
In PBS (0.09% sodium azide) | |
RP11-295F4.3/SCZD5/TA4/TRAR4 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
319100 | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-7 ug/mL) The optimal working dilution should be determined by the end user. | |
TAAR6 | |
TAAR6 | |
A synthetic peptide corresponding to 16 amino acid at cytoplasmic domain of human TAAR6. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only