Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACR1, Rabbit, DyLight 488, Polyclonal Antibody, Abnova™

Catalog No. 89117488 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-488 Supplier Abnova Corporation Supplier No. PAB15188
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TACR1.

This gene belongs to a gene family of tachykinin receptors. These tachykinin receptors are characterized by interactions with G proteins and contain seven hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin substance P, also referred to as neurokinin 1. The encoded protein is also involved in the mediation of phosphatidylinositol metabolism of substance P. [provided by RefSeq

Specifications

Antigen TACR1
Applications Flow Cytometry, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 488
Description Rabbit polyclonal antibody raised against synthetic peptide of TACR1.
Dilution Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene TACR1
Gene Alias NK1R/NKIR/SPR/TAC1R
Gene Symbols TACR1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human TACR1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6869
Test Specificity This is specific to human NK-1. This product may be used for Western blotting where a band is seen at ∼53 KDa representing human NK-1. The actual molecular weight may vary depending upon the tissue used.
Target Species Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.