Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACR1 Rabbit anti-Dog, Guinea pig, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115518 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-518 Supplier Abnova Corporation Supplier No. PAB12008
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Tacr1.

Specifications

Antigen TACR1
Applications Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Tacr1.
Dilution Immunofluorescence (1:50) Immunohistochemistry (Frozen sections) (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene Tacr1
Gene Alias Tac1r
Gene Symbols Tacr1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with TG) corresponding to C-terminus of rat Tacr1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24807
Test Specificity This antibody is specific to the NK-1 receptor.
Target Species Canine, Guinea Pig, Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.