Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACR3 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114360 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-360 Supplier Abnova Corporation Supplier No. PAB0710
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Tacr3.

Sequence: ASTTSSFISSPYTSVDEYS

Specifications

Antigen TACR3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Tacr3.
Dilution Western Blot (1:500-1:2000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Tacr3
Gene Alias Nkr/Nmkr/Tac3r
Gene Symbols Tacr3
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 434-452 of rat Tacr3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24808
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.