Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Tau, AEP-cleaved (N368), Polyclonal,

Catalog No. ABN1703MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1703MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1703MI Supplier MilliporeSigma™ Supplier No. ABN1703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Tau, AEP-cleaved (N368) in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Microtubule-associated protein Tau (UniProt P10636; also known as Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau) is encoded by the MAPT (also known as TAU, MAPT1, MTBTL) gene (Gene ID 4137) in human. Neurofibrillary tangles (NFTs), composed of truncated and hyperphosphorylated Tau, are a common feature of aging-related neurodegenerative diseases, including Alzheimer's disease (AD). In addition to caspases-mediated cleavage at Asp421, Tau can also be cleaved at Asn255 and Asn368 by the lysosomal cysteine proteinase asparagine endopeptidase (AEP). AEP is upregulated in aged brain, as well as n human AD brain and tau P301S–transgenic mice with synaptic pathology and behavioral impairments. AEP-mediated tau cleavage abolishes tau microtubule assembly function, induces tau aggregation and triggers neurodegeneration. AEP knockout or functional blockage in tau P301S mice leads to a reduction of tau hyperphosphorylation, protection against memory loss, and a reduction in tau P301S–triggered pathological and behavioral defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau, AEP-cleaved (N368)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P10636
Gene Symbols MAPT; MAPTL; MTBT1; TAU
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human Tau, AEP-cleaved (N368).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005901
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.