Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-TDP-43, Polyclonal,

Catalog No. ABN895MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN895MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN895MI Supplier MilliporeSigma™ Supplier No. ABN895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TDP-43 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

TAR DNA-binding protein 43 (TDP-43) belongs to the hnRNP protein family and plays an important role in transcription, pre-mRNA splicing, mRNA stability and mRNA transport. It is involved in splicing of the apolipoprotein A-II and cystic fibrosis transmembrane gene. This protein is highly expressed in the pancreas, placenta, lung, genital tract and spleen. Mutations in TDP-43 have been associated with amyotrophic lateral sclerosis, frontotemporal dementia, Parkinson's disease and Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDP-43
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.
Gene Accession No. Q13148
Gene Symbols TARDBP; TDP43
Host Species Rabbit
Immunogen TDP43 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human TDP43.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.