Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD10, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-846 Supplier Abnova Corporation Supplier No. PAB22139

Rabbit polyclonal antibody raised against recombinant TDRD10.

Sequence: LLVTSIVPKTPFFWAMHVTEALHQNMQALFSTLAQAEEQQPYLEGSTVMRGTRCLAEYHLGDYGHAWNRCWVLDRVDTWAV

Specifications

Antigen TDRD10
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description tudor domain containing 10
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene TDRD10
Gene Alias DKFZp434M202
Gene Symbols TDRD10
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human TDRD10.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126668
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.