Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF2, Rabbit, DyLight 649, Polyclonal Antibody, Abnova™

Catalog No. 89138039 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-039 Supplier Abnova Corporation Supplier No. PAB15118
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant TERF2.

Specifications

Antigen TERF2
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 649
Description Rabbit polyclonal antibody raised against recombinant TERF2.
Dilution ChIP (1:2000-1:5000) Immunofluorescence (1:2000-1:5000) Western Blot (1:2000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Host Species Rabbit
Immunogen Recombinant protein corresponding to TERF2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TRF2 This antibody is useful for Western blot, where a band at ∼56 KDa is seen.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.