Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TF Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118689 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-689 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-689 Supplier Abnova Corporation Supplier No. PAB16876
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native TF.

This gene encodes a glycoprotein with an approximate molecular weight of 76.5kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq

Specifications

Antigen TF
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native TF.
Dilution Western Blot (1:50000) ELISA (1:150000-1:300000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.01M PBS, pH 7.4.
Gene TF
Gene Alias DKFZp781D0156/PRO1557/PRO2086
Gene Symbols TF
Host Species Rabbit
Immunogen Native purified TF from human transferrin.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Human
Content And Storage Store at -20°C. After reconstitution with 500μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.