Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGS1 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117163 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-163 Supplier Abnova Corporation Supplier No. PAB14587

Specifications

Antigen TGS1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In serum (50% glycerol, 0.025% sodium azide)
Gene Symbols Tgs1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 408-423 of mouse Tgs1.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 116940
Target Species Human, Mouse
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.