Learn More
TH (phospho S40), Rabbit anti-Rat, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | TH |
| Applications | Immunofluorescence, Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic phosphopeptide of Th. |
| Dilution | Immunohistochemistry (Frozen sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
| Formulation | In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) |
| Gene | Th |
| Gene Alias | The |
| Gene Symbols | Th |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.