Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THPO Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118066 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-066 Supplier Abnova Corporation Supplier No. PAB16175
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant THPO.

Megakaryocytopoiesis is the cellular development process that leads to platelet production. The protein encoded by this gene is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene. [provided by RefSeq

Specifications

Antigen THPO
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant THPO.
Dilution ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS.
Gene THPO
Gene Alias MGC163194/MGDF/MKCSF/ML/MPLLG/TPO
Gene Symbols THPO
Host Species Rabbit
Immunogen Recombinant protein corresponding to human THPO.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7066
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.