Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thromboxane B2 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137652 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-652 Supplier Abnova Corporation Supplier No. PAB13666

Rabbit polyclonal antibody raised against Thromboxane B2.

Specifications

Antigen Thromboxane B2
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Thromboxane B2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Rabbit
Immunogen Thromboxane B2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antisera is specific to Thromboxane B2 (TXB2). HAPTEN REACTIVITY: TXB2 100% PGD2 <1% PGE2 <1% PGF2alpha <1% 6,keto-PGF1alpha <1%
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.