Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-TICAM2, Polyclonal, Abnova™

Catalog No. 89117206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-206 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-206 Supplier Abnova Corporation Supplier No. PAB14639

Specifications

Antigen TICAM2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Symbols TICAM2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 87-103 of human TICAM2.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 353376
Target Species Canine, Chimpanzee, Equine, Human, Macaque, Mouse, Rat
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.