Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TICAM2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134377 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-377 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-377 Supplier Abnova Corporation Supplier No. PAB9740
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TICAM2.

TIRP is a Toll/interleukin-1 receptor (IL1R; MIM 147810) (TIR) domain-containing adaptor protein involved in Toll receptor signaling (see TLR4; MIM 603030).[supplied by OMIM

Specifications

Antigen TICAM2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TICAM2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene TICAM2
Gene Alias MGC129876/MGC129877/TICAM-2/TIRAP3/TIRP/TRAM
Gene Symbols TICAM2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human TICAM2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353376
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.