Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-TIRP, Polyclonal,

Catalog No. ABF318MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF318MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF318MI Supplier MilliporeSigma™ Supplier No. ABF318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TIRP in Human samples, and it is validated for Immunocytochemistry, Western Blotting

Toll/interleukin-1 receptor domain-containing protein (TIRP), also known as TIR domain-containing adapter molecule 2 (TICAM2), also known as Putative NF-kappa-B-activating protein 502, TRIF-related adapter molecule (TRAM), and Toll-like receptor adaptor protein 3 (TIRAP3), is an adaptor protein containing one TIR domain, and is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) serving as an important link between the innate and adaptive immune responses. TIRP is expressed in spleen, prostate, testis, uterus, small intestine, colon, peripheral blood leukocytes, heart, placenta, lung, liver, skeletal muscle, and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIRP
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.
Gene Accession No. Q86XR7
Gene Symbols TICAM2; TIRAP3; TIRP; TRAM
Host Species Rabbit
Immunogen TIRP antibody was raised against a peptide corresponding to 16 amino acids at the C-terminus of human TIRP.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.