Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLK1 Rabbit anti-Chicken, Chimpanzee, Dog, Human, Mouse, Orangutan, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114715 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-715 Supplier Abnova Corporation Supplier No. PAB10238
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TLK1.

The Tousled-like kinases, first described in Arabidopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.[supplied by OMIM

Specifications

Antigen TLK1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TLK1.
Dilution ELISA (1:10000-1:50000) Western Blot (1:200-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene TLK1
Gene Alias KIAA0137/PKU-beta
Gene Symbols TLK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 737-751 of human TLK1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9874
Test Specificity This antibody reacts with both phosphorylated and non-phosphorylated forms of Tlk-1 at the S743 residue (isoform 1) of the protein. This antibody will react with all isoforms of Tlk-1.
Target Species Canine, Chicken, Chimpanzee, Human, Mouse, Orangutan, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.