Learn More
TLR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TLR1.
Supplier: Abnova Corporation PAB9724
Description
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is ubiquitously expressed, and at higher levels than other TLR genes. Different length transcripts presumably resulting from use of alternative polyadenylation site, and/or from alternative splicing, have been noted for this gene. [provided by RefSeq
Specifications
TLR1 | |
Polyclonal | |
toll-like receptor 1 | |
TLR1 | |
TLR1 | |
A synthetic peptide corresponding to internal region of human TLR1. | |
RUO | |
7096 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
CD281, DKFZp547I0610, DKFZp564I0682, KIAA0012, MGC104956, MGC126311, MGC126312, TIL, rsc786 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only